Sodium bisulfite, powder ACS

Supplier: Thermo Scientific Chemicals
Warning

41944-1000 AC41944-0010 AC41944-0025 AC41944-0050
TS41944-1000EA 40 USD
TS41944-1000 TS41944-0010 TS41944-0025 TS41944-0050
Sodium bisulfite, powder ACS
Sodium bisulphite
Formula: HNaO₃S
Melting Pt: 150 °C (dec.)
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7631-90-5
EINECS: 231-548-0
Merck Index: 12,08731

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodimetric ≥58.5 % (SO2)
Chloride (Cl) ≤200 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR