Orthophosphoric acid 85+% in water ACS

Supplier: Thermo Scientific Chemicals
Danger

42404-5000 424040025 424040100
TS42404-5000EA 124 USD
TS42404-5000 TS424040025 TS424040100
Orthophosphoric acid 85+% in water ACS
Orthophosphoric acid
Formula: H₃PO₄
Melting Pt: 21 °C
Density: 1.68 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear liquid
Color scale ≤10 APHA
Titration with NaOH ≥85.0 %
Chloride (Cl) ≤3 ppm
Nitrate (NO3) ≤5 ppm
Sulfate (SO4) ≤30 ppm
Antimony (Sb) ≤20 ppm
Arsenic (As) ≤1 ppm
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤30 ppm
Magnesium (Mg) ≤20 ppm
Manganese (Mn) ≤0.5 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤250 ppm
Heavy metals (as Pb) ≤10 ppm
Volatile acids (CH3COOH) ≤0.001 %
Insoluble matter ≤0.001 %
Reducing substances Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR