Sodium hydroxide 98%, soluble, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Caustic soda

45058-0025 450580100 450580250
TS45058-0025EA 74 USD
TS45058-0025 TS450580100 TS450580250
Sodium hydroxide 98%, soluble, pure
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance White Flakes
Titration with HCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR