Sodium hydroxide, micropearls for analysis

Supplier: Thermo Scientific Chemicals
Danger

38304-0025 383040010 383042500
TS38304-0025EA 183 USD
TS38304-0025 TS383040010 TS383042500
Sodium hydroxide, micropearls for analysis
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance White Pearls
Titration with HCl ≥99.0 %
Sodium carbonate (Na2CO3) ≤0.4 %
Sodium sulfate (Na2SO4) ≤80 ppm
Sodium chloride (NaCl) ≤200 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR