Potassium iodate 99.4+% ACS

Supplier: Thermo Scientific Chemicals
Danger

41824-5000 AC41824-0025 AC41824-1000
TS41824-5000EA 262 USD
TS41824-5000 TS41824-0025 TS41824-1000
Potassium iodate 99.4+% ACS
Potassium iodate
Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560 °C
Density: 3.93 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 13,07726

Order Now

Specification Test Results

Appearance White Powder or crystalline powder
Titration Iodometric 99.4 to 100.4 %
pH 5.0 to 8.0 (5% soln. at 25°C)
Chloride and Bromide ≤0.01 % (as Cl)
Iodide (I) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR