Sodium iodide, anhydrous ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
41957-1000
AC41957-0025
AC41957-5000
TS41957-1000EA
83
USD
TS41957-1000
TS41957-0025
TS41957-5000
Sodium iodide, anhydrous ACS
Sodium iodide
Formula:
NaI MW: 149.89 g/mol Boiling Pt: 1300 °C (760 mmHg) Melting Pt: 661 °C Density: 3.66 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White Solid |
pH | 6.0 to 9.0 (5 % solution at 25.0°C) |
Titration with KIO3 | ≥99.5 % |
Chloride and Bromide | ≤0.01 % (as Cl) |
Iodate (IO3) | ≤3 ppm |
Phosphate (PO4) | ≤0.001 % |
Sulfate (SO4) | ≤0.005 % |
Barium (Ba) | ≤0.002 % |
Calcium (Ca) | ≤0.002 % |
Iron (Fe) | ≤5 ppm |
Magnesium (Mg) | ≤0.001 % |
Potassium (K) | ≤0.01 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...