Sodium iodide, anhydrous 99+%, pure

Supplier: Thermo Scientific Chemicals
Warning

20318-0010 20318-0025 20318-2500 AC20318-0050 AC20318-0100
TS20318-0010EA 544 USD
TS20318-0010 TS20318-0025 TS20318-2500 TS20318-0050 TS20318-0100
Sodium iodide, anhydrous 99+%, pure
Sodium iodide
Formula: NaI
Boiling Pt: 1300 °C (760 mmHg)
Melting Pt: 661 °C
Density: 3.66 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥99.0 % (on dried substance)
Loss on drying ≤3 % (1 g, 105°C, 3 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR