Sodium cyanotrihydroborate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium cyanoborohydride

16855-0100 16855-0500 16855-2500
TS16855-0100EA 81 USD
TS16855-0100 TS16855-0500 TS16855-2500
Sodium cyanotrihydroborate 95%
Sodium cyanotrihydroborate
Formula: CH₃BNNa
MW: 62.84 g/mol
Melting Pt: >242 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003516
CAS Number: 25895-60-7
EINECS: 247-317-2
UN: 3134
ADR: 4.3,II
Merck Index: 13,08680

Order Now

Specification Test Results

Appearance White Powder
Titration ≥94.0 %
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR