Sodium fluoride ≥99%, powder ACS

Supplier: Thermo Scientific Chemicals
Danger

42432-5000 AC42432-0050
TS42432-5000EA 146 USD
TS42432-5000 TS42432-0050
Sodium fluoride ≥99%, powder ACS
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
UN: 1690
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Loss on drying ≤0.3 % (150°C)
Titration ≥99 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤300 ppm
Sodium fluosilicate ≤0.1 % (Na2SiF6)
Sulfite (SO2) ≤50 ppm
Iron (Fe) ≤30 ppm
Potassium (K) ≤200 ppm
Heavy metals (as Pb) ≤30 ppm
Titrable acid ≤0.03 meq/g
Titrable base ≤0.01 meq/g
Insoluble matter ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR