Lithium fluoride 99.98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

19338-1000 AC19338-0050 AC19338-0250
TS19338-1000EA 549 USD
TS19338-1000 TS19338-0050 TS19338-0250
Lithium fluoride 99.98%, Extra Pure
Lithium fluoride
Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1676 °C (1013 hPa)
Melting Pt: 845 °C
Density: 2.63 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,05553

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR