Potassium fluoride, anhydrous 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
20135-5000
AC20135-0010
AC20135-0250
AC20135-2500
TS20135-5000EA
187
USD
TS20135-5000
TS20135-0010
TS20135-0250
TS20135-2500
Potassium fluoride, anhydrous 99%, Extra Pure
Potassium fluoride
Formula:
KF MW: 58.1 g/mol Boiling Pt: 1505 °C (1013 hPa) Melting Pt: 860 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III Merck Index: 13,07714 |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or chunks |
Titration | ≥98.5 % |
Chloride (Cl) | ≤500 ppm |
Sulfate (SO4) | ≤1000 ppm |
Iron (Fe) | ≤50 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...