Potassium dichromate 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

AC42411-0025 AC42411-5000
TS42411-0500EA 83.18 USD
TS42411-0500 TS42411-0025 TS42411-5000 TS424110500
Potassium dichromate 99+% ACS
Potassium dichromate
Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3087
ADR: 5.1 II
REACH: 01-2119454792-32
Merck Index: 12,07790

Order Now

Specification Test Results

Appearance Orange Crystals
Titration Iodometric ≥99.0 %
Loss on drying ≤0.05 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤50 ppm
Calcium (Ca) ≤30 ppm
Iron (Fe) ≤10 ppm
Sodium (Na) ≤200 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR