Lithium chloride 99%, extra pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
19337-0010
AC19337-0050
TS19337-0010EA
260
USD
TS19337-0010
TS19337-0050
Lithium chloride 99%, extra pure
Lithium chloride
Formula:
LiCl Boiling Pt: 1382 °C (760 mmHg) Melting Pt: 605 °C Density: 2.06 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 |
Specification Test Results
Appearance | White Crystalline powder or crystals or granules |
Titration Argentometric | ≥98.5 % |
Loss on drying | ≤0.5 % (400°C, 4 h) |
Insoluble matter | ≤0.01 % (in acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...