Potassium chloride 99+% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Potassiumchloride

19677-0010 AC19677-0050 AC19677-0250
TS19677-0010EA 154 USD
TS19677-0010 TS19677-0050 TS19677-0250
Potassium chloride 99+% for analysis
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Argentometric 99.0 to 100.5 % (on dried substance)
Loss on drying ≤0.5 % (1 g, 105°C, 3 h)
Acidity/alkalinity complies
Bromide (Br) ≤1000 ppm
Iodide (I) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Aluminium (Al) ≤1 ppm
Arsenic (As) ≤0.5 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤2 ppm
Magnesium (Mg) ≤50 ppm
Sodium (Na) ≤700 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR