Sodium carbonate, anhydrous 99.5%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
12367-0100
AC12367-0010
AC12367-0025
TS12367-0100EA
606
USD
TS12367-0100
TS12367-0010
TS12367-0025
Sodium carbonate, anhydrous 99.5%, Extra Pure
Sodium carbonate
Formula:
Na₂CO₃ Boiling Pt: 1600 °C (760 mmHg) Melting Pt: 851 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 REACH: 01-2119485498-19 Merck Index: 14,08596 |
|
Specification Test Results
Appearance | White Granular powder |
Loss on drying | ≤1 % (300°C, 2 hrs) |
Titration with HCl | 99.5 to 100.5 % (On dried substance) |
Chloride (Cl) | ≤125 ppm |
Sulfate (SO4) | ≤250 ppm |
Arsenic (As) | ≤5 ppm |
Iron (Fe) | ≤50 ppm |
Heavy metals (as Pb) | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...