Sodium bromide ACS
Supplier: Thermo Scientific Chemicals
21267-5000
AC21267-0025
AC21267-1000
TS21267-5000EA
113
USD
TS21267-5000
TS21267-0025
TS21267-1000
Sodium bromide ACS
Sodium bromide
Formula:
NaBr MW: 102.89 g/mol Boiling Pt: 1396 Melting Pt: 753 Storage Temperature: Ambient |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 Merck Index: 13,08666 |
Specification Test Results
Appearance | Colorless to white Powder or solid |
Titration Argentometric | ≥99.0 % |
pH | 5.0 to 8.8 |
Chloride (Cl) | ≤0.2 % |
Bromate (BrO3) | ≤0.001 % |
Sulfate (SO4) | ≤0.002 % |
Barium (Ba) | ≤0.002 % |
Calcium (Ca) | ≤0.002 % |
Iron (Fe) | ≤0.0005 % |
Magnesium (Mg) | ≤0.001 % |
Potassium (K) | ≤0.1 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...