Potassium bromide 99+% for IR spectrophotometry
Supplier: Thermo Scientific Chemicals
|
Warning
|
20639-0250
20639-1000
20639-5000
TS20639-0250EA
169
USD
TS20639-0250
TS20639-1000
TS20639-5000
Potassium bromide 99+% for IR spectrophotometry
Potassium bromide
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 Melting Pt: 730 Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 13,07701 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥99.0 % |
Additional info | Application test: Passes test |
Absorbance | ≤0.015 (other: 420-4000 1/cm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...