Lithium tetrahydroborate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
20681-0100
20681-0500
AC20681-0050
TS20681-0100EA
169
USD
TS20681-0100
TS20681-0500
TS20681-0050
Lithium tetrahydroborate 95%
Lithium tetrahydroborate
Formula:
H₄BLi Boiling Pt: 66 °C (1013 hPa) Melting Pt: 280 °C Density: 0.66 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011088 CAS Number: 16949-15-8 EINECS: 241-021-7 UN: 1413 ADR: 4.3,I Merck Index: 13,05547 |
Specification Test Results
Appearance | White Powder |
Assay | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...