1,4-Dioxane 99+% stabilized ACS, meets analytical specifications of Reagent Ph. Eur.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

40882-0010 408820025 408820100 408825000
TS40882-0010EA 126 USD
TS40882-0010 TS408820025 TS408820100 TS408825000
1,4-Dioxane 99+% stabilized ACS, meets analytical specifications of Reagent Ph. Eur.
1,4-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1 °C (1013 hPa)
Melting Pt: 12 °C
Density: 1.034 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
UN: 1165
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
Density (20°C) 1.032 to 1.036 g/ml
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 5 to 30 ppm BHT
Residue after evaporation ≤0.005 %
Carbonyl (as HCHO) ≤0.01 %
Titrable acid ≤0.0016 meq/g
Peroxides ≤0.005 % (H2O2)
Freezing point ≥11.0°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR