cis-Cyclooctene 90% stabilised, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Z)-Cyclooctene

15486-0010 15486-0025 15486-2500
TS15486-0010EA 209 USD
TS15486-0010 TS15486-0025 TS15486-2500
cis-Cyclooctene 90% stabilised, Technical Grade
cis-Cyclooctene
Formula: C₈H₁₄
Boiling Pt: 184…191 °C (7 mmHg)
Melting Pt: 18…20 °C
Density: 0.84 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00001753
CAS Number: 931-87-3
EINECS: 220-846-6
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Slightly hazy colorless to white to light yellow Liquid or waxy solid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4640 (20°C, 589 nm)
GC ≥88 %
Stabilizer Stabilized with BHT


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR