Cyclohexane ACS

Supplier: Thermo Scientific Chemicals
Danger

44628-0050 44628-0010
TS44628-0050EA 161 USD
TS44628-0050 TS44628-0010
Cyclohexane ACS
Cyclohexane
Formula: C₆H₁₂
Boiling Pt: 81 °C (760 mmHg)
Melting Pt: 6.5 °C
Density: 0.77 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
UN: 1145
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 %
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Substance darkened by H2SO4 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR