Cyclohexane, extra dry 99.5%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

32683-0010 326830025 326831000
TS32683-0010EA 126 USD
TS32683-0010 TS326830025 TS326831000
Cyclohexane, extra dry 99.5%, AcroSeal™
Cyclohexane
Formula: C₆H₁₂
Boiling Pt: 81 °C (760 mmHg)
Melting Pt: 6.5 °C
Density: 0.77 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.4 %
Residue after evaporation ≤0.0005 %
Acidity ≤0.001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR