Cyclopentanone 99+%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
11153-0100
11153-0010
11153-0025
11153-2500
TS11153-0100EA
1331
USD
TS11153-0100
TS11153-0010
TS11153-0025
TS11153-2500
Cyclopentanone 99+%, pure
Cyclopentanone
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 130…131 °C (760 mmHg) Melting Pt: –51 °C Density: 0.95 g/cm³ Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001409 CAS Number: 120-92-3 EINECS: 204-435-9 UN: 2245 ADR: 3,III Merck Index: 13,02771 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4360 to 1.4380 (20°C, 589 nm) |
GC | ≥88 % |
Stabilizer | 0.01 % alpha-Tocopherol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...