Benzyl benzoate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzoic acid benzyl ester

10586-0010 10586-0051 10586-2500
TS10586-0010EA 111 USD
TS10586-0010 TS10586-0051 TS10586-2500
Benzyl benzoate 99+%
Benzyl benzoate
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 323 °C (760 mmHg)
Melting Pt: 18…20 °C
Density: 1.118 g/cm³
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00003075
CAS Number: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9,III
Merck Index: 13,01129

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5670 to 1.5700 (20°C, 589 nm)
GC ≥99.0 %
Specific gravity (20/20°C) 1.118 to 1.122
Acid value ≤0.3 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR