Cyclohexylbenzene 99.5% for analysis
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phenylcyclohexane
11129-5000
11129-1000
TS11129-5000EA
190
USD
TS11129-5000
TS11129-1000
Cyclohexylbenzene 99.5% for analysis
Cyclohexylbenzene
Formula:
C₁₂H₁₆ MW: 160.26 g/mol Boiling Pt: 239 °C (1013 hPa) Melting Pt: 135…152 °C Density: 0.95 g/cm³ Flash Pt: 174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001451 CAS Number: 827-52-1 EINECS: 201-069-1 UN: 3082 ADR: 9,III Merck Index: 13,02350 |
Specification Test Results
Appearance | Colorless to white Powder or crystals |
Infrared spectrum | Conforms |
Water | 7.5 to 9.0 % (K.F.) |
Heavy metals | ≤5 ppm |
Sulfated ash | ≤0.1 % |
Readily carbonizable substance | Passes test |
Titration with NaOH | ≥99.4 % |
Chloride (Cl) | ≤10 ppm |
Oxalate (C2O4) | ≤500 ppm |
Sulfate (SO4) | ≤100 ppm |
Iron (Fe) | ≤3 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...