1,4-Diethylbenzene 95%

Supplier: Thermo Scientific Chemicals
Danger

11415-0250 11415-0050
TS11415-0250EA 193 USD
TS11415-0250 TS11415-0050
1,4-Diethylbenzene 95%
1,4-Diethylbenzene
Formula: C₁₀H₁₄
Boiling Pt: 247 °C (760 mmHg)
Melting Pt: 17…20 °C
Density: 0.82 g/cm³
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00009264
CAS Number: 105-05-5
EINECS: 228-222-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4340 (20°C, 589 nm)
GC ≥94 %
UV (10 % in water)
Titration with NaOH ≥98.5 %
Membrane filtration test (10 % in water) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR