2,4-Xylenol 99%

Supplier: Thermo Scientific Chemicals
Danger

40845-5000 40845-1000 40845-0050 40845-0250
TS40845-5000EA 215 USD
TS40845-5000 TS40845-1000 TS40845-0050 TS40845-0250
2,4-Xylenol 99%
2,4-Xylenol
Formula: C₈H₁₀O
Boiling Pt: 211 °C
Melting Pt: 21…23 °C
Density: 1.027 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00002233
CAS Number: 105-67-9
EINECS: 203-321-6
UN: 2928
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5375 to 1.5405 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR