α,α'-Dichloro-p-xylene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Bis(chloromethyl)benzene, p-Xylene dichloride, p-Xylylene dichloride

14766-1000 14766-5000 14766-0250
TS14766-1000EA 92 USD
TS14766-1000 TS14766-5000 TS14766-0250
α,α'-Dichloro-p-xylene 98%
α,α'-Dichloro-p-xylene
Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 254 °C (760 mmHg)
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00000920
CAS Number: 623-25-6
EINECS: 210-782-7
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline solid
Infrared spectrum Conforms
Melting point 98 to 102 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR