Anisole, extra dry 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl phenyl ether, Methoxybenzene

42925-1000 42925-0010
TS42925-1000EA 123 USD
TS42925-1000 TS42925-0010
Anisole, extra dry 99%, AcroSeal™
Anisole
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 154 °C (760 mmHg)
Melting Pt: –37 °C
Density: 0.99 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
UN: 2222
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d32206d4bb9dbd12360d11d39b31237d54a8dcf8&vid=286a07a5e34a1ec55ff6339785af95d9a4326825&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.5 %
Residue after evaporation ≤0.005 %
UV 1 cm cell vs HPLC-grade water
Non-volatile matter ≤0.0003 %
Free acid (CH3COOH) ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR