Anisole 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

15392-0010 15392-2500 15392-0025 15392-0050
TS15392-0010EA 167 USD
TS15392-0010 TS15392-2500 TS15392-0025 TS15392-0050
Anisole 99%, pure
Anisole
Formula: C₇H₈O
Boiling Pt: 154 °C (760 mmHg)
Melting Pt: –37 °C
Density: 0.99 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
EINECS: 202-876-1
UN: 2222
ADR: 3,III
Merck Index: 13,00672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d32206d4bb9dbd12360d11d39b31237d54a8dcf8&vid=286a07a5e34a1ec55ff6339785af95d9a4326825&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5190 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR