Chlorobenzene, extra dry 99.8%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Warning
|
39697-0010
396971000
TS39697-0010EA
146.29
USD
TS39697-0010
TS396971000
Chlorobenzene, extra dry 99.8%, AcroSeal™
Chlorobenzene
Formula:
C₆H₅Cl Boiling Pt: 131 °C (760 mmHg) Melting Pt: –45 °C Density: 1.1075 g/cm³ Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000530 CAS Number: 108-90-7 UN: 1134 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.005 % (Coulometric) |
GC | ≥99.8 % |
Residue after evaporation | ≤0.001 % |
Filtered to 0.2 micrometer | Filtered |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...