3,5-Dinitrosalicylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxy-3,5-dinitrobenzoic acid, 3,5-Dinitro-2-hydroxybenzoic acid
15644-0010
15644-1000
TS15644-0010EA
587
USD
TS15644-0010
TS15644-1000
3,5-Dinitrosalicylic acid 98%
3,5-Dinitrosalicylic acid
Formula:
C₇H₄N₂O₇ MW: 228.12 g/mol Melting Pt: 170…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007104 CAS Number: 609-99-4 EINECS: 210-204-3 |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
Melting point | 170 to 174 °C |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...