Benzoic acid 98+% ACS

Supplier: Thermo Scientific Chemicals
Warning

42347-0250 42347-5000 42347-0020 42347-0100
TS42347-0250EA 42 USD
TS42347-0250 TS42347-5000 TS42347-0020 TS42347-0100
Benzoic acid 98+% ACS
Benzoic acid
Formula: C₇H₆O₂
Boiling Pt: 131…132 °C (760 mmHg)
Melting Pt: –103 °C
Density: 0.75 g/cm³
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 217-730-2
UN: 1993
ADR: 3,III
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4225 to 1.4275 (20°C, 589 nm)
GC ≥98.0 %
Chlorine compounds (as Cl) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as S) ≤0.002 %
Insoluble matter ≤0.005 % (in methanol)
KMnO4-reducing substance Passes test
Freezing point 122°C to 123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR