3,5-Dihydroxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

42565-0050 42565-0010 42565-0250
TS42565-0050EA 263 USD
TS42565-0050 TS42565-0010 TS42565-0250
3,5-Dihydroxybenzaldehyde 98%
3,5-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
Melting Pt: 153…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00016611
CAS Number: 26153-38-8

Order Now

Specification Test Results

Appearance White to gray to tan Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR