Salicylaldehyde 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxybenzaldehyde

13260-0010 13260-0050 13260-1000 13260-2500
TS13260-0010EA 174 USD
TS13260-0010 TS13260-0050 TS13260-1000 TS13260-2500
Salicylaldehyde 99%
Salicylaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 197 °C (760 mmHg)
Melting Pt: –7 °C
Density: 1.16 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00003317
CAS Number: 90-02-8
EINECS: 201-961-0
UN: 2810
ADR: 6.1,III
Merck Index: 13,08405

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5715 to 1.5745 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤200 APHA
Phenol ≤1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR