p-Anisaldehyde 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Methoxybenzaldehyde
10480-0050
10480-1000
10480-0025
10480-5000
TS10480-0050EA
55
USD
TS10480-0050
TS10480-1000
TS10480-0025
TS10480-5000
p-Anisaldehyde 99+%
p-Anisaldehyde
Formula:
C₈H₈O₂ Boiling Pt: 248…249 °C (760 mmHg) Melting Pt: –1 °C Density: 1.121 g/cm³ Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003385 CAS Number: 123-11-5 EINECS: 204-602-6 Merck Index: 13,00666 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5710 to 1.5740 (20°C, 589 nm) |
GC | ≥99.0 % |
Relative density | (D20/4°C) 1.120 to 1.124 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...