Bromobenzene 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenylbromide, Phenyl bromide

10668-0010 10668-0025 10668-2500 10668-0100
TS10668-0010EA 144 USD
TS10668-0010 TS10668-0025 TS10668-2500 TS10668-0100
Bromobenzene 99%, pure
Bromobenzene
Formula: C₆H₅Br
MW: 157.01 g/mol
Boiling Pt: 156 °C (760 mmHg)
Melting Pt: –31 °C
Density: 1.49 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000055
CAS Number: 108-86-1
EINECS: 203-623-8
UN: 2514
ADR: 3,III
Merck Index: 13,01390

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5580 to 1.5600 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR