1,4-Dibromobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11273-0025
11273-0010
11273-2500
TS11273-0025EA
361
USD
TS11273-0025
TS11273-0010
TS11273-2500
1,4-Dibromobenzene 99%
1,4-Dibromobenzene
Formula:
C₆H₄Br₂ Boiling Pt: 219 °C (1013 hPa) Melting Pt: 86…89 °C Density: 1.84 g/cm³ Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000089 CAS Number: 106-37-6 EINECS: 203-390-2 UN: 3077 ADR: 9,III Merck Index: 13,03042 |
Specification Test Results
Appearance | Colorless to off-white Crystals |
Infrared spectrum | Conforms |
Melting point | 86 to 89 °C |
GC | ≥98.5 % |
Chloride and Bromide | ≤0.01 % (as Cl) |
Iodate (IO3) | ≤3 ppm |
Phosphate (PO4) | ≤0.001 % |
Sulfate (SO4) | ≤0.005 % |
Barium (Ba) | ≤0.002 % |
Calcium (Ca) | ≤0.002 % |
Iron (Fe) | ≤3 ppm |
Magnesium (Mg) | ≤0.001 % |
Sodium (Na) | ≤0.005 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...