2-Methyl-2-butene, anhydrous 99+% for molecular biology
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Trimethylethylene, Isoamylene, Isopentene
12649-2500
12649-0250
TS12649-2500EA
297
USD
TS12649-2500
TS12649-0250
2-Methyl-2-butene, anhydrous 99+% for molecular biology
2-Methyl-2-butene
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 310 °C (1013 hPa) Melting Pt: 153 °C Density: 0.66 g/cm³ Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009276 CAS Number: 513-35-9 EINECS: 201-069-1 UN: 2460 ADR: 3,II REACH: 01-2119457026-42-0000 Merck Index: 13,02350 |
Specification Test Results
Appearance | White Crystalline solid |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Titration with NaOH | ≥99 % |
Dnase,Rnase,Protease activity | none detected |
Carboniz. sub. by hot H2SO4 | (as per ACS) passes test |
Chloride (Cl) | ≤10 ppm |
Oxalate (C2O4) | ≤500 ppm |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...