Vinyl acetate 99+% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid vinyl ester

14084-0025 14084-0010 14084-0100 14084-0250
TS14084-0025EA 75 USD
TS14084-0025 TS14084-0010 TS14084-0100 TS14084-0250
Vinyl acetate 99+% stabilized
Vinyl acetate
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 72…73 °C (760 mmHg)
Melting Pt: –93 °C
Density: 0.93 g/cm³
Flash Pt: –8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008713
CAS Number: 108-05-4
EINECS: 203-545-4
UN: 1301
ADR: 3,II
Merck Index: 12,10130

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3940 to 1.3960 (20°C, 589 nm)
Water ≤0.05 %
GC ≥99.0 %
Stabilizer 3 to 15 ppm Hydroquinone
Acidity (CH3COOH) ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR