2-Methylbutane 99+%, extra_pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
iso-Pentane
12647-0010
126470025
126470250
TS12647-0010EA
133
USD
TS12647-0010
TS126470025
TS126470250
2-Methylbutane 99+%, extra_pure
2-Methylbutane
Formula:
CH₃CH₂CH(CH₃)₂ MW: 72.15 g/mol Boiling Pt: 28 Melting Pt: -160 Density: 0.62 g/cm³ Flash Pt: –51 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009338 CAS Number: 78-78-4 EINECS: 201-142-8 UN: 1265 ADR: 3 I |
|
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3530 to 1.3550 (20°C, 589 nm) |
GC | ≥99.0 % |
Residue after evaporation | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...