2,6-Dimethyl-4-heptanone, technical , remainder mainly 4,6-dimethyl-2-heptanone
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diisobutyl Ketone
14944-0200
14944-0010
14944-0025
TS14944-0200EA
572
USD
TS14944-0200
TS14944-0010
TS14944-0025
2,6-Dimethyl-4-heptanone, technical , remainder mainly 4,6-dimethyl-2-heptanone
2,6-Dimethyl-4-heptanone
Formula:
C₉H₁₈O Boiling Pt: 165 °C (1013 hPa) Melting Pt: –46 °C Density: 0.809 g/cm³ Flash Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008940 CAS Number: 108-83-8 EINECS: 203-620-1 UN: 1157 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4100 to 1.4180 (20°C, 589 nm) |
Water | ≤0.2 % (K.F.) |
GC | ≥15 % (4.6-Dimethyl-2-heptanone) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...