n-Tetradecane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetradecane

17413-0025 17413-1000 17413-5000
TS17413-0025EA 772 USD
TS17413-0025 TS17413-1000 TS17413-5000
n-Tetradecane 99%
n-Tetradecane
Formula: H₃C(CH₂)₁₂CH₃
MW: 198.39 g/mol
Boiling Pt: 252…254 °C (760 mmHg)
Melting Pt: 5.8 °C
Density: 0.767 g/cm³
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS Number: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR