n-Octane 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

12937-0025 12937-5000 129370020 129370250
TS12937-0025EA 674 USD
TS12937-0025 TS12937-5000 TS129370020 TS129370250
n-Octane 99+%, Extra Pure
n-Octane
Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 125…127 °C (760 mmHg)
Melting Pt: –57 °C
Density: 0.708 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,II
Merck Index: 13,06782
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68adf890d68728825d4a2fa4b032700b93f710b8&vid=b00a2925902bc425c80f8ebb3f0e7b456c800ae3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3965 to 1.3985 (20°C, 589 nm)
Density (15°C) 0.706 to 0.710 kg/l
GC ≥99.0 %
Residue after evaporation ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR