n-Dodecane 99%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dodecane
11759-0025
11759-5000
11759-2500
117591000
117590100
117590250
TS11759-0025EA
838
USD
TS11759-0025
TS11759-5000
TS11759-2500
TS117591000
TS117590100
TS117590250
n-Dodecane 99%, pure
n-Dodecane
Formula:
H₃C(CH₂)₁₀CH₃ MW: 170.34 g/mol Boiling Pt: 215…217 °C (760 mmHg) Melting Pt: –12 °C Density: 0.753 g/cm³ Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008969 CAS Number: 112-40-3 EINECS: 203-967-9 |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4210 to 1.4230 (20 °C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤10 APHA |
Heavy metals (ICP-OES) | ≤5 ppm (as Pb) |
Titrable acid | ≤0.0008 meq/g |
Insoluble matter | ≤0.01 % |
Reducing sugars | Passes test |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...