Sodium dodecyl sulfate (SDS) 85%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
22614-0025
AC22614-1000
AC22614-5000
TS22614-0025EA
237
USD
TS22614-0025
TS22614-1000
TS22614-5000
Sodium dodecyl sulfate (SDS) 85%, pure
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 206 °C Flash Pt: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 Merck Index: 14,08636 |
|
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
GC | ≥80 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...