Sodium decyl sulfate for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
40622-0250
40622-1000
AC40622-0050
TS40622-0250EA
569
USD
TS40622-0250
TS40622-1000
TS40622-0050
Sodium decyl sulfate for HPLC
Sodium decyl sulphate
Formula:
C₁₀H₂₁NaO₄S Storage Temperature: Ambient |
MDL Number:
MFCD00041881 CAS Number: 142-87-0 EINECS: 205-568-5 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Water | ≤1.5 % (K.F.) |
Solubility | (25 % in water at 25°C) Clear colorless to light yellow |
UV | at 280 nm A: ≤0.01 |
Titration after Ion exchange | ≥98.5 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...