Glycerine 99.5+% for spectroscopy

Supplier: Thermo Scientific Chemicals

18469-0025 18469-5000
TS18469-0025EA 435 USD
TS18469-0025 TS18469-5000
Glycerine 99.5+% for spectroscopy
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
Boiling Pt: 290 °C (760 mmHg)
Melting Pt: 18 °C
Density: 1.261 g/cm³
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.5 % (Coulometric)
GC ≥99.50 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR