1,3-Propanediol 98%

Supplier: Thermo Scientific Chemicals

13140-0010 13140-0025 13140-0100 13140-2500
TS13140-0010EA 324 USD
TS13140-0010 TS13140-0025 TS13140-0100 TS13140-2500
1,3-Propanediol 98%
1,3-Propanediol
Formula: HO(CH₂)₃OH
Boiling Pt: 214 °C (760 mmHg)
Melting Pt: –32 °C
Density: 1.052 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00002949
CAS Number: 504-63-2
EINECS: 207-997-3
Merck Index: 12,09845

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥97.5 %
Color scale ≤20 APHA
UV at 400 nm A: ≤0.009
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR