Ethylene glycol, anhydrous 99.8%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Warning
|
43381-0010
43381-1000
TS43381-0010EA
122.23
USD
TS43381-0010
TS43381-1000
Ethylene glycol, anhydrous 99.8%, AcroSeal™
Ethylene glycol
Formula:
HOCH₂CH₂OH Boiling Pt: 196…198 °C (760 mmHg) Melting Pt: –13 °C Density: 1.113 g/cm³ Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002885 CAS Number: 107-21-1 |
Specification Test Results
Appearance | Clear colorless Viscous liquid |
Infrared spectrum | Conforms |
Water | ≤0.01 % |
GC | ≥99.75 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...